Product Details

SNP ID
rs2718107
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:129198818 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTTTGTGTAAGTACACTCTGATGTT[A/C]ACACAAAGATGACATTGCCTGATGA
Phenotype
MIM: 601500
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SMO PubMed Links

Gene Details

Gene
SMO
Gene Name
smoothened, frizzled class receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005631.4 Intron NP_005622.1
XM_011516522.2 Intron XP_011514824.1

View Full Product Details