Product Details

SNP ID
rs3112329
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:135671135 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATCATTCTCCTTCAAGGTGCAACA[C/T]TGGTAAAAGTAGGATTTTGCAGGCC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C7orf73 PubMed Links

Gene Details

Gene
C7orf73
Gene Name
chromosome 7 open reading frame 73
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130929.1 Intron NP_001124401.1

View Full Product Details