Product Details
- SNP ID
-
hCV26520319
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:35215685 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TAAATATTTTTTCTAGGCAAAAAAA[A/G]GGGCATCATTACCTTCTTTGATCTT
- Phenotype
-
MIM: 606061
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TBX20
PubMed Links
Gene Details
- Gene
- TBX20
- Gene Name
- T-box 20
View Full Product Details