Product Details
- SNP ID
-
rs481247
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:58933395 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CTAAATGATCCAAGCCCAGCTGTCC[C/T]TCCTGATTTTACTCCCTCCAGGCTT
- Phenotype
-
MIM: 614761
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
GLYATL1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs142020980] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- GLYATL1
- Gene Name
- glycine-N-acyltransferase like 1
- Gene
- LOC283194
- Gene Name
- uncharacterized LOC283194
There are no transcripts associated with this gene.
View Full Product Details