Product Details

SNP ID
rs1009712
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:59642493 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAAAGGTCTCAATAAGGAAGATGG[A/C]CAGCTGGTCCACATCTTTTTGCCCT
Phenotype
MIM: 614660
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PATL1 PubMed Links

Gene Details

Gene
PATL1
Gene Name
PAT1 homolog 1, processing body mRNA decay factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152716.2 Intron NP_689929.2
XM_005273833.4 Intron XP_005273890.1
XM_005273834.1 Intron XP_005273891.1
XM_011544811.2 Intron XP_011543113.1
XM_017017339.1 Intron XP_016872828.1

View Full Product Details