Product Details

SNP ID
rs1683213
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:78191391 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGACGCCCCTCAGGCTGTCATGCCA[C/G]TAGGGCGAGGGGTATGAGGCAAGGG
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
KCTD21 PubMed Links

Gene Details

Gene
KCTD21
Gene Name
potassium channel tetramerization domain containing 21
There are no transcripts associated with this gene.

Gene
USP35
Gene Name
ubiquitin specific peptidase 35
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020798.2 Intron NP_065849.1
XM_011545187.2 Intron XP_011543489.1
XM_011545188.2 Intron XP_011543490.1
XM_017018052.1 Intron XP_016873541.1
XM_017018053.1 Intron XP_016873542.1

View Full Product Details