Product Details

SNP ID
rs11906796
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:36065993 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCAGGACACGATCCCTGACTTTGAA[A/T]ACCTTTTTTTGCGGGTTTTGGGGAA
Phenotype
MIM: 602879
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
EPB41L1 PubMed Links

Gene Details

Gene
EPB41L1
Gene Name
erythrocyte membrane protein band 4.1 like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001258329.1 Intron NP_001245258.1
NM_001258330.1 Intron NP_001245259.1
NM_001258331.1 Intron NP_001245260.1
NM_012156.2 Intron NP_036288.2
NM_177996.2 Intron NP_818932.1
XM_011528667.1 Intron XP_011526969.1
XM_011528671.1 Intron XP_011526973.1
XM_011528673.1 Intron XP_011526975.1
XM_011528677.1 Intron XP_011526979.1
XM_011528681.1 Intron XP_011526983.1
XM_011528682.1 Intron XP_011526984.1
XM_011528683.1 Intron XP_011526985.1
XM_011528685.1 Intron XP_011526987.1
XM_017027711.1 Intron XP_016883200.1
XM_017027712.1 Intron XP_016883201.1
XM_017027713.1 Intron XP_016883202.1
XM_017027714.1 Intron XP_016883203.1
XM_017027715.1 Intron XP_016883204.1
XM_017027716.1 Intron XP_016883205.1
XM_017027717.1 Intron XP_016883206.1
XM_017027718.1 Intron XP_016883207.1
XM_017027719.1 Intron XP_016883208.1
XM_017027720.1 Intron XP_016883209.1

View Full Product Details