Product Details

SNP ID
rs2234733
Assay Type
DME
NCBI dbSNP Submissions
NA
Location
Chr.6:52752852 on Build GRCh38
Set Membership
DME Validated Inventoried
Context Sequence [VIC/FAM]
CCCAAAACACTGAACAGCTTCACTT[A/C]CTTTTTCAAAGGCAGGGAAGTAGCG
Phenotype
MIM: 138360
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
GSTA2 PubMed Links

Gene Details

Gene
GSTA2
Gene Name
glutathione S-transferase alpha 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000846.4 Intron NP_000837.3
XM_011514532.2 Intron XP_011512834.1
Gene
GSTA7P
Gene Name
glutathione S-transferase alpha 7, pseudogene
There are no transcripts associated with this gene.

View Full Product Details