Product Details

SNP ID
rs35975455
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:62168861 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTCAGGCATAAACTAGAACCAAA[C/G]AATGGAGATCTGGGAAACCCTAGAA
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
RTKN2 PubMed Links

Gene Details

Gene
RTKN2
Gene Name
rhotekin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282941.1 Intron NP_001269870.1
NM_145307.3 Intron NP_660350.2
XM_011539456.2 Intron XP_011537758.1
XM_011539457.2 Intron XP_011537759.1
XM_011539460.2 Intron XP_011537762.1
XM_017015842.1 Intron XP_016871331.1
XM_017015843.1 Intron XP_016871332.1
XM_017015844.1 Intron XP_016871333.1

View Full Product Details