Product Details

SNP ID
rs136147
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:36256843 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GATTCCTCTGTGTTTGAATGGGAAA[G/T]TGGTACCCATCTCCTGGCCTGTGTG
Phenotype
MIM: 603743
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
APOL1 PubMed Links

Gene Details

Gene
APOL1
Gene Name
apolipoprotein L1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136540.1 Intron NP_001130012.1
NM_001136541.1 Intron NP_001130013.1
NM_003661.3 Intron NP_003652.2
NM_145343.2 Intron NP_663318.1
XM_005261796.3 Intron XP_005261853.1
XM_011530478.2 Intron XP_011528780.1

View Full Product Details