Product Details

SNP ID
rs3767316
Assay Type
Functionally tested
NCBI dbSNP Submissions
42
Location
Chr.1:95239497 on Build GRCh38
Set Membership
HapMap JSNP
Context Sequence [VIC/FAM]
TGAGTTTTTAAAAGTATGTAATTTT[A/G]TTGCCACATTAAATACCAGAAGAAT
Phenotype
MIM: 615875
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC101928118 PubMed Links

Gene Details

Gene
LOC101928118
Gene Name
uncharacterized LOC101928118
There are no transcripts associated with this gene.

Gene
RWDD3
Gene Name
RWD domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001128142.1 Intron NP_001121614.1
NM_001199682.1 Intron NP_001186611.1
NM_001278247.1 Intron NP_001265176.1
NM_001278248.1 Intron NP_001265177.1
NM_015485.4 Intron NP_056300.2
Gene
TMEM56-RWDD3
Gene Name
TMEM56-RWDD3 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199691.1 Intron NP_001186620.1

View Full Product Details