Product Details

SNP ID
rs12725780
Assay Type
Functionally tested
NCBI dbSNP Submissions
29
Location
Chr.1:233329195 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACTTTGCAAACCGAGGCCTTCTGC[A/G]GATTATATGGAACCCATGACCGTCT
Phenotype
MIM: 614793
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KIAA1804 PubMed Links

Gene Details

Gene
KIAA1804
Gene Name
mixed lineage kinase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032435.2 Intron NP_115811.2
XM_011544305.2 Intron XP_011542607.1

View Full Product Details