Product Details
- SNP ID
-
rs1800298
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:80101841 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCTCGTCCGCCCGTTGTTCAGCGAG[C/G]GAGGCTCTGCGCGTGCCGCAGCTGA
- Phenotype
-
MIM: 613799
MIM: 606800
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
CCDC40
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs80020206] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CCDC40
- Gene Name
- coiled-coil domain containing 40
There are no transcripts associated with this gene.
- Gene
- GAA
- Gene Name
- glucosidase alpha, acid
- Gene
- MIR1268B
- Gene Name
- microRNA 1268b
There are no transcripts associated with this gene.
View Full Product Details