Product Details

SNP ID
rs2156994
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:73800639 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGTCTCCTCAGTATCATCCCTTC[A/G]AGTTTCACCAGAAAGATGTTATCAG
Phenotype
MIM: 300181
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TSIX PubMed Links

Gene Details

Gene
TSIX
Gene Name
TSIX transcript, XIST antisense RNA
There are no transcripts associated with this gene.

View Full Product Details