Product Details

SNP ID
rs386584
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:11822223 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CACAGGGAGGTGGTCACCAGAATGC[A/G]GTTGTGTAATAAATATATGTGAGTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF440 PubMed Links

Gene Details

Gene
ZNF440
Gene Name
zinc finger protein 440
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152357.2 Intron NP_689570.2
XM_005259731.3 Intron XP_005259788.1
XM_017026253.1 Intron XP_016881742.1
XM_017026254.1 Intron XP_016881743.1

View Full Product Details