Product Details

SNP ID
rs4264508
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:15472833 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTTTCACAGAATTAGACAAAACTA[C/T]CCTGAAATTCGCAAGGAACCAAAAA
Phenotype
MIM: 608199 MIM: 616561
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PGLYRP2 PubMed Links

Gene Details

Gene
PGLYRP2
Gene Name
peptidoglycan recognition protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_052890.3 Intron NP_443122.3
XM_006722633.3 Intron XP_006722696.1
Gene
RASAL3
Gene Name
RAS protein activator like 3
There are no transcripts associated with this gene.

View Full Product Details