Product Details

SNP ID
rs35749757
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:51821667 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGGCGCGGCCGTGGGGAAGCCATA[G/T]GCGGTGGGCGGTGCCGGCGCGGGCA
Phenotype
MIM: 600702
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
FIGNL2 PubMed Links

Gene Details

Gene
FIGNL2
Gene Name
fidgetin like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001013690.4 1161 Silent Mutation GCA,GCC A249A NP_001013712.4
XM_005268870.3 1161 Silent Mutation GCA,GCC A249A XP_005268927.1
XM_017019298.1 1161 Silent Mutation GCA,GCC A363A XP_016874787.1
XM_017019299.1 1161 Silent Mutation GCA,GCC A347A XP_016874788.1
Gene
SCN8A
Gene Name
sodium voltage-gated channel alpha subunit 8
There are no transcripts associated with this gene.

View Full Product Details