Product Details

SNP ID
rs1355237
Assay Type
Functionally tested
NCBI dbSNP Submissions
10
Location
Chr.1:149899725 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGAACACAGGGGCTTCGGCCCAA[C/T]CACAAGTACGCAGTTGCACATGCCC
Phenotype
MIM: 613181 MIM: 185860
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BOLA1 PubMed Links

Gene Details

Gene
BOLA1
Gene Name
bolA family member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321025.1 144 Intron NP_001307954.1
NM_001321026.1 144 UTR 5 NP_001307955.1
NM_016074.4 144 Intron NP_057158.1
XM_006711348.3 144 UTR 5 XP_006711411.1
Gene
SV2A
Gene Name
synaptic vesicle glycoprotein 2A
There are no transcripts associated with this gene.

View Full Product Details