Product Details

SNP ID
rs15931
Assay Type
Functionally tested
NCBI dbSNP Submissions
32
Location
Chr.1:149884800 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTGAACCAATGAGGCTTCTCTTGGC[C/T]TCATTTGTAAAAAGGAGATTAACTC
Phenotype
MIM: 615014 MIM: 602797 MIM: 601831
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HIST2H2AB PubMed Links

Gene Details

Gene
HIST2H2AB
Gene Name
histone cluster 2, H2ab
There are no transcripts associated with this gene.

Gene
HIST2H2AC
Gene Name
histone cluster 2, H2ac
There are no transcripts associated with this gene.

Gene
HIST2H2BE
Gene Name
histone cluster 2, H2be
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003528.2 1883 UTR 3 NP_003519.1

View Full Product Details