Product Details

SNP ID
rs535537
Assay Type
Functionally tested
NCBI dbSNP Submissions
63
Location
Chr.1:109658504 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTTCTTTGTATCCTTGATTTTGCTC[A/G]TGTCTGGGTCCAGAGCCTGCCAGGT
Phenotype
MIM: 138380 MIM: 138333
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GSTM2 PubMed Links

Gene Details

Gene
GSTM2
Gene Name
glutathione S-transferase mu 2 (muscle)
There are no transcripts associated with this gene.

Gene
GSTM4
Gene Name
glutathione S-transferase mu 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000850.4 Intron NP_000841.1
NM_147148.2 Intron NP_671489.1
XM_017001085.1 Intron XP_016856574.1
XM_017001086.1 Intron XP_016856575.1
XM_017001087.1 Intron XP_016856576.1
XM_017001088.1 Intron XP_016856577.1

View Full Product Details