Product Details

SNP ID
hCV26634463
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:43187004 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATTGCTAGAACAAAGATTTATGCA[A/T]TTTAAATATTCCTTTATTATAAAAC
Phenotype
MIM: 607089
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CCNDBP1 PubMed Links

Gene Details

Gene
CCNDBP1
Gene Name
cyclin D1 binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012142.4 Intron NP_036274.3
Gene
TMEM62
Gene Name
transmembrane protein 62
There are no transcripts associated with this gene.

View Full Product Details