Product Details

SNP ID
rs2608492
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:52436705 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCTTTCAGTTTGCTCATTGTTTTCC[A/T]TTCCTTTTGCTGTCTGCTGTTAAAC
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ZNF534 PubMed Links

Gene Details

Gene
ZNF534
Gene Name
zinc finger protein 534
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143938.1 Intron NP_001137410.1
NM_001143939.1 Intron NP_001137411.1
NM_001291368.1 Intron NP_001278297.1
NM_001291369.1 Intron NP_001278298.1

View Full Product Details