Product Details

SNP ID
rs2732021
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:25059730 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGGGCATCCGCAGGTAAATGTCTG[A/C]CATCACACGCACATTAGGAACATTC
Phenotype
MIM: 605436
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SNORD116-1 PubMed Links

Gene Details

Gene
SNORD116-1
Gene Name
small nucleolar RNA, C/D box 116-1
There are no transcripts associated with this gene.

Gene
SNORD116-2
Gene Name
small nucleolar RNA, C/D box 116-2
There are no transcripts associated with this gene.

Gene
SNORD116-3
Gene Name
small nucleolar RNA, C/D box 116-3
There are no transcripts associated with this gene.

Gene
SNORD116-4
Gene Name
small nucleolar RNA, C/D box 116-4
There are no transcripts associated with this gene.

Gene
SNORD116-5
Gene Name
small nucleolar RNA, C/D box 116-5
There are no transcripts associated with this gene.

Gene
SNORD116-6
Gene Name
small nucleolar RNA, C/D box 116-6
There are no transcripts associated with this gene.

Gene
SNORD116-7
Gene Name
small nucleolar RNA, C/D box 116-7
There are no transcripts associated with this gene.

Gene
SNORD116@
Gene Name
small nucleolar RNA, C/D box 116 cluster
There are no transcripts associated with this gene.

View Full Product Details