Product Details

SNP ID
rs11784544
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:11346994 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGATTGCAATATGCAGCGGAGTTT[C/G]AGAACCAGCGCCTTGGACCCAGCTT
Phenotype
MIM: 615174
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
TDH PubMed Links

Gene Details

Gene
TDH
Gene Name
L-threonine dehydrogenase (pseudogene)
There are no transcripts associated with this gene.

View Full Product Details