Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001076683.1 | 2640 | UTR 3 | NP_001070151.1 | ||
NM_001076684.2 | 2640 | UTR 3 | NP_001070152.1 | ||
NM_014233.3 | 2640 | UTR 3 | NP_055048.1 | ||
XM_006722059.3 | 2640 | UTR 3 | XP_006722122.1 | ||
XM_006722060.2 | 2640 | Intron | XP_006722123.1 | ||
XM_006722061.2 | 2640 | Intron | XP_006722124.1 | ||
XM_017024995.1 | 2640 | UTR 3 | XP_016880484.1 | ||
XM_017024996.1 | 2640 | UTR 3 | XP_016880485.1 | ||
XM_017024997.1 | 2640 | UTR 3 | XP_016880486.1 | ||
XM_017024998.1 | 2640 | UTR 3 | XP_016880487.1 | ||
XM_017024999.1 | 2640 | UTR 3 | XP_016880488.1 | ||
XM_017025000.1 | 2640 | UTR 3 | XP_016880489.1 | ||
XM_017025001.1 | 2640 | UTR 3 | XP_016880490.1 | ||
XM_017025002.1 | 2640 | UTR 3 | XP_016880491.1 | ||
XM_017025003.1 | 2640 | UTR 3 | XP_016880492.1 | ||
XM_017025004.1 | 2640 | UTR 3 | XP_016880493.1 |