Product Details

SNP ID
rs2924052
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:97086560 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCTTACTGTGTGTCAGGCACTGTT[C/T]TAAGTGTTTTAATCTATTAACTCAC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAHD2B PubMed Links

Gene Details

Gene
FAHD2B
Gene Name
fumarylacetoacetate hydrolase domain containing 2B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320848.1 Intron NP_001307777.1
NM_001320849.1 Intron NP_001307778.1
NM_199336.2 Intron NP_955368.1
XM_011510745.2 Intron XP_011509047.1
XM_011510746.2 Intron XP_011509048.1
XM_011510747.2 Intron XP_011509049.1
XM_011510748.2 Intron XP_011509050.1
XM_011510750.2 Intron XP_011509052.1
XM_017003470.1 Intron XP_016858959.1
XM_017003471.1 Intron XP_016858960.1
XM_017003472.1 Intron XP_016858961.1
XM_017003473.1 Intron XP_016858962.1
Gene
LOC653924
Gene Name
glycerol-3-phosphate acyltransferase 2, mitochondrial pseudogene
There are no transcripts associated with this gene.

View Full Product Details