Product Details

SNP ID
rs4441523
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:96191368 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATACAGGGGGTTCATTATACTCTTC[C/T]ACTTCTGTATCTTTTGTATCTATTT
Phenotype
MIM: 616712
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
STARD7 PubMed Links

Gene Details

Gene
STARD7
Gene Name
StAR related lipid transfer domain containing 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020151.3 Intron NP_064536.2

View Full Product Details