Product Details
- SNP ID
-
rs4392265
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:112283438 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCAAATAAATAAATAATCAAGTTCC[G/A]AGTAGTTTCTAAAGAGAGTAAACAT
- Phenotype
-
- Polymorphism
- G/A, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ZC3H6
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs143986709] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ZC3H6
- Gene Name
- zinc finger CCCH-type containing 6
View Full Product Details