Product Details

SNP ID
rs2867298
Assay Type
Functionally tested
NCBI dbSNP Submissions
29
Location
Chr.1:150701551 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CATGTCCAATTTAAGCTACCAACAT[A/G]ATATCTCTGAGTGGGGAGTTGGGAA
Phenotype
MIM: 612208 MIM: 609824
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GOLPH3L PubMed Links

Gene Details

Gene
GOLPH3L
Gene Name
golgi phosphoprotein 3 like
There are no transcripts associated with this gene.

Gene
HORMAD1
Gene Name
HORMA domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199829.1 Intron NP_001186758.1
NM_032132.4 Intron NP_115508.2
XM_006711570.2 Intron XP_006711633.1
XM_006711571.2 Intron XP_006711634.1
XM_011510053.2 Intron XP_011508355.1
XM_011510054.2 Intron XP_011508356.1
XM_011510055.2 Intron XP_011508357.1
XM_011510056.2 Intron XP_011508358.1

View Full Product Details