Product Details

SNP ID
rs698914
Assay Type
Functionally tested
NCBI dbSNP Submissions
46
Location
Chr.1:150326292 on Build GRCh38
Set Membership
HapMap JSNP
Context Sequence [VIC/FAM]
CACCTCATTGTTGCCTCTTTGTAGA[A/G]GTTTCTCTGCAATGCACTTGTAGAT
Phenotype
MIM: 607301
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PRPF3 PubMed Links

Gene Details

Gene
PRPF3
Gene Name
pre-mRNA processing factor 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004698.2 Intron NP_004689.1
XM_011510129.1 Intron XP_011508431.1
XM_011510130.2 Intron XP_011508432.1
XM_011510131.2 Intron XP_011508433.1
XM_011510132.2 Intron XP_011508434.1
XM_017002790.1 Intron XP_016858279.1
XM_017002791.1 Intron XP_016858280.1

View Full Product Details