Product Details
- SNP ID
-
rs572238427
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:108740576 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCTTAGAAATGAAAGGACATTTCTA[C/T]ATGCATGCTGGTTCTCTGCTTTTGA
- Phenotype
-
MIM: 601181
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
RANBP2
PubMed Links
Gene Details
- Gene
- RANBP2
- Gene Name
- RAN binding protein 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_006267.4 |
959 |
Silent Mutation |
TAC,TAT |
Y290Y |
NP_006258.3 |
XM_005264002.2 |
959 |
Silent Mutation |
TAC,TAT |
Y290Y |
XP_005264059.1 |
XM_005264003.2 |
959 |
Silent Mutation |
TAC,TAT |
Y290Y |
XP_005264060.1 |
XM_005264004.2 |
959 |
Silent Mutation |
TAC,TAT |
Y290Y |
XP_005264061.1 |
XM_005264005.4 |
959 |
Silent Mutation |
TAC,TAT |
Y264Y |
XP_005264062.1 |
XM_005264007.2 |
959 |
Silent Mutation |
TAC,TAT |
Y290Y |
XP_005264064.1 |
XM_011511575.2 |
959 |
Silent Mutation |
TAC,TAT |
Y289Y |
XP_011509877.1 |
XM_011511576.2 |
959 |
Silent Mutation |
TAC,TAT |
Y290Y |
XP_011509878.1 |
XM_011511578.2 |
959 |
Silent Mutation |
TAC,TAT |
Y289Y |
XP_011509880.1 |
XM_017004623.1 |
959 |
Intron |
|
|
XP_016860112.1 |
XM_017004624.1 |
959 |
Silent Mutation |
TAC,TAT |
Y290Y |
XP_016860113.1 |
XM_017004625.1 |
959 |
Intron |
|
|
XP_016860114.1 |
View Full Product Details