Product Details
- SNP ID
-
hCV26674310
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:108731625 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTAAAAAGTGTGTTAAAACCTTTCT[C/G]AGCATCTACTGTCTTATTAGGCATT
- Phenotype
-
MIM: 601181
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
RANBP2
PubMed Links
Gene Details
- Gene
- RANBP2
- Gene Name
- RAN binding protein 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_006267.4 |
|
Intron |
|
|
NP_006258.3 |
View Full Product Details