Product Details

SNP ID
rs2216526
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:95811348 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAATCAAATCATGTACTAGGCTGAA[G/A]TTAAATCAAATACCATTTACTGAGT
Phenotype
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
LINC00342 PubMed Links

Gene Details

Gene
LINC00342
Gene Name
long intergenic non-protein coding RNA 342
There are no transcripts associated with this gene.

View Full Product Details