Product Details

SNP ID
rs2394385
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.10:73688922 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAACAACACTGTAATAAGAGCACA[A/G]CTGCCTCTGCCTCTGCCTCCTCTGC
Phenotype
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
AGAP5 PubMed Links

Gene Details

Gene
AGAP5
Gene Name
ArfGAP with GTPase domain, ankyrin repeat and PH domain 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001144000.1 Intron NP_001137472.1

View Full Product Details