Product Details
- SNP ID
-
rs12461523
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:39517580 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ATATGAGCGAAGCCCTGAAGGAAGG[A/C]GAGAAGGAGTTGTGTGGATGCTGTG
- Phenotype
-
MIM: 602768
MIM: 607919
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
DLL3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs73037270] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- DLL3
- Gene Name
- delta like canonical Notch ligand 3
There are no transcripts associated with this gene.
- Gene
- SELV
- Gene Name
- selenoprotein V
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_182704.1 |
|
Intron |
|
|
NP_874363.1 |
View Full Product Details