Product Details
- SNP ID
-
rs2233708
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:39605460 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAACAGGAGGCAGCTCTCATTGATC[A/T]GGCCTCAGTAGTCCGTCAGGGTCCA
- Phenotype
-
MIM: 608717
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
LGALS13
PubMed Links
Gene Details
- Gene
- LGALS13
- Gene Name
- galectin 13
View Full Product Details