Product Details
- SNP ID
-
rs2233710
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:39607112 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAGTGTATCTAATACGTTAACTTGT[A/G]TAACTAGGAATTTTCTTGGGGAATG
- Phenotype
-
MIM: 608717
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
LGALS13
PubMed Links
Gene Details
- Gene
- LGALS13
- Gene Name
- galectin 13
View Full Product Details