Product Details

SNP ID
rs2376481
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:26975566 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGTCTACGTTATGGCTAGTTAACAC[A/G]CTTTAAAAGACTCATGTATATATTA
Phenotype
MIM: 600233
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GABRG3 PubMed Links

Gene Details

Gene
GABRG3
Gene Name
gamma-aminobutyric acid type A receptor gamma3 subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270873.1 Intron NP_001257802.1
NM_033223.4 Intron NP_150092.2
XM_011521430.2 Intron XP_011519732.1
XM_011521431.2 Intron XP_011519733.1
XM_017022058.1 Intron XP_016877547.1
XM_017022059.1 Intron XP_016877548.1
XM_017022060.1 Intron XP_016877549.1

View Full Product Details