Product Details

SNP ID
rs12004983
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:33751747 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGATGGCTTATTGTTCATGAGAAAC[A/G]TGTACTGTTTTTAACCAGATGTGTT
Phenotype
MIM: 613578
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PRSS3 PubMed Links

Gene Details

Gene
PRSS3
Gene Name
protease, serine 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001197097.2 Intron NP_001184026.2
NM_001197098.1 Intron NP_001184027.1
NM_002771.3 Intron NP_002762.2
NM_007343.3 Intron NP_031369.2
XM_011517965.1 Intron XP_011516267.1

View Full Product Details