Product Details

SNP ID
hCV26698976
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:32982020 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCCTAACATTGAGATTGTTTAGTT[C/T]GCCACATGTTTATTGATGGTCTACT
Phenotype
MIM: 615470 MIM: 610884
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CEP89 PubMed Links

Gene Details

Gene
CEP89
Gene Name
centrosomal protein 89
There are no transcripts associated with this gene.

Gene
FAAP24
Gene Name
Fanconi anemia core complex associated protein 24
There are no transcripts associated with this gene.

Gene
RHPN2
Gene Name
rhophilin Rho GTPase binding protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033103.4 Intron NP_149094.3

View Full Product Details