Product Details

SNP ID
rs2405220
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:75414204 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GATGAGTTCCAAGTTACTATTGATT[A/G]TAGACATAAAATTTATTATCAAAGG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SMIM21 PubMed Links

Gene Details

Gene
SMIM21
Gene Name
small integral membrane protein 21
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001037331.3 Intron NP_001032408.1
NM_001303482.1 Intron NP_001290411.1
XM_011525928.1 Intron XP_011524230.1

View Full Product Details