Product Details

SNP ID
rs4502555
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:113852985 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCTTGGATTTAGTACTCTATAAGT[A/G]AGCAGAAAGAGATTCAATAAATAGT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GRAMD1C PubMed Links

Gene Details

Gene
GRAMD1C
Gene Name
GRAM domain containing 1C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001172105.1 Intron NP_001165576.1
NM_017577.4 Intron NP_060047.3
XM_005247546.2 Intron XP_005247603.1
XM_005247547.1 Intron XP_005247604.1
XM_011512930.1 Intron XP_011511232.1
XM_011512931.1 Intron XP_011511233.1
XM_017006646.1 Intron XP_016862135.1
XM_017006647.1 Intron XP_016862136.1
XM_017006648.1 Intron XP_016862137.1
XM_017006649.1 Intron XP_016862138.1

View Full Product Details