Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001163508.1 | Intron | NP_001156980.1 | |||
NM_001163509.1 | Intron | NP_001156981.1 | |||
NM_015604.3 | Intron | NP_056419.2 | |||
NM_181340.2 | Intron | NP_851937.1 | |||
NM_181341.2 | Intron | NP_851938.2 | |||
XM_017021201.1 | Intron | XP_016876690.1 | |||
XM_017021202.1 | Intron | XP_016876691.1 | |||
XM_017021203.1 | Intron | XP_016876692.1 | |||
XM_017021204.1 | Intron | XP_016876693.1 | |||
XM_017021205.1 | Intron | XP_016876694.1 | |||
XM_017021206.1 | Intron | XP_016876695.1 | |||
XM_017021207.1 | Intron | XP_016876696.1 | |||
XM_017021208.1 | Intron | XP_016876697.1 | |||
XM_017021209.1 | Intron | XP_016876698.1 | |||
XM_017021210.1 | Intron | XP_016876699.1 | |||
XM_017021211.1 | Intron | XP_016876700.1 | |||
XM_017021212.1 | Intron | XP_016876701.1 | |||
XM_017021213.1 | Intron | XP_016876702.1 | |||
XM_017021214.1 | Intron | XP_016876703.1 | |||
XM_017021215.1 | Intron | XP_016876704.1 |