Product Details

SNP ID
rs4132325
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:112212887 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCTCCCCACCCGCACACACACACA[C/T]TGACACTCAAAGTCATCAAGAAACT
Phenotype
MIM: 607679 MIM: 605465
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DOCK4 PubMed Links

Gene Details

Gene
DOCK4
Gene Name
dedicator of cytokinesis 4
There are no transcripts associated with this gene.

Gene
ZNF277
Gene Name
zinc finger protein 277
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021994.2 Intron NP_068834.2
XM_005250129.3 Intron XP_005250186.1
XM_011515768.2 Intron XP_011514070.1
XM_017011720.1 Intron XP_016867209.1

View Full Product Details