Product Details

SNP ID
rs2518042
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:4998660 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAGGCTTGTACTTGAGCCCTGGCT[C/T]GTTGAGGATCATCTCCAGCAGCCTG
Phenotype
MIM: 600450
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AKR1C2 PubMed Links

Gene Details

Gene
AKR1C2
Gene Name
aldo-keto reductase family 1, member C2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135241.2 Intron NP_001128713.1
NM_001321027.1 Intron NP_001307956.1
NM_001354.5 Intron NP_001345.1
NM_205845.2 Intron NP_995317.1

View Full Product Details