Product Details

SNP ID
rs7922
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:125236454 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGCTAATGAAAAGCAGTAAACAGCC[A/G]CTTAGGCTATAGCAGTTTCAACTCC
Phenotype
MIM: 138120 MIM: 605962
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HSPA5 PubMed Links

Gene Details

Gene
HSPA5
Gene Name
heat shock protein family A (Hsp70) member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005347.4 2364 UTR 3 NP_005338.1
Gene
RABEPK
Gene Name
Rab9 effector protein with kelch motifs
There are no transcripts associated with this gene.

View Full Product Details