Product Details

SNP ID
rs3935732
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:67340411 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTCTATTCTACTAAATGACTGATA[A/G]CTTTACAATGAGATTTGTTTTATTT
Phenotype
MIM: 604450
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PSMD12 PubMed Links

Gene Details

Gene
PSMD12
Gene Name
proteasome 26S subunit, non-ATPase 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001316341.1 2195 UTR 3 NP_001303270.1
NM_002816.4 2195 UTR 3 NP_002807.1
NM_174871.3 2195 UTR 3 NP_777360.1

View Full Product Details