Product Details

SNP ID
rs3020885
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:15580320 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTGTTCAACTCTATTTCAGATTG[C/T]TCTTATATAAATTCAATCCAGCAAG
Phenotype
MIM: 604596
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CDRT1 PubMed Links

Gene Details

Gene
CDRT1
Gene Name
CMT1A duplicated region transcript 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282540.1 Intron NP_001269469.1
NM_006382.3 Intron NP_006373.2

View Full Product Details