Product Details

SNP ID
rs4652419
Assay Type
Functionally Tested
NCBI dbSNP Submissions
42
Location
Chr.1:179592548 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTATCCCACTATTGGTTTTTTTTT[A/T]AATCTTTTTTCGTGGGTTTGCCCCC
Phenotype
Polymorphism
A/T, Transversion Substitution
Allele Nomenclature
Literature Links
TDRD5 PubMed Links
Additional Information
For this assay, SNP(s) [rs76241720] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
TDRD5
Gene Name
tudor domain containing 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199085.1 292 Intron NP_001186014.1
NM_001199089.1 292 Intron NP_001186018.1
NM_001199091.1 292 UTR 5 NP_001186020.1
NM_001199092.1 292 Intron NP_001186021.1
NM_173533.3 292 Intron NP_775804.2
XM_005244934.1 292 Intron XP_005244991.1
XM_005244935.4 292 Intron XP_005244992.1
XM_017000473.1 292 Intron XP_016855962.1

View Full Product Details