Product Details

SNP ID
rs4654938
Assay Type
Functionally tested
NCBI dbSNP Submissions
63
Location
Chr.1:21459488 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACGTTTTCCAAGAACACACAGGAGG[G/T]CTATGGGTAGAGGAAGGTGTCTGTC
Phenotype
MIM: 612992
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
NBPF3 PubMed Links

Gene Details

Gene
NBPF3
Gene Name
neuroblastoma breakpoint family member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256416.2 Intron NP_001243345.1
NM_001256417.2 Intron NP_001243346.1
NM_032264.4 Intron NP_115640.1
XM_006710957.1 Intron XP_006711020.1
XM_006710958.1 Intron XP_006711021.1
XM_006710959.2 Intron XP_006711022.1
XM_011542276.1 Intron XP_011540578.1
XM_011542277.1 Intron XP_011540579.1
XM_011542278.1 Intron XP_011540580.1
XM_011542281.1 Intron XP_011540583.1
XM_017002498.1 Intron XP_016857987.1
XM_017002499.1 Intron XP_016857988.1
XM_017002500.1 Intron XP_016857989.1
XM_017002501.1 Intron XP_016857990.1
XM_017002502.1 Intron XP_016857991.1

View Full Product Details